Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYH4
Entrez Gene ID
4622
Gene Name
myosin heavy chain 4
Gene Aliases
MYH2B, MyHC-2B, MyHC-IIb
Chromosome Location
Chr.17: 10443058 - 10469559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017533.2 NP_060003.2 2 - 3 77 75
GenBank mRNA AF111783.2 - 2 - 3 75

View Full Product Details