The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288725.1 | NP_001275654.1 | 5 - 6 | 714 | 108 |
NM_001288726.1 | NP_001275655.1 | 5 - 6 | 711 | 108 | |
NM_019040.4 | NP_061913.3 | 5 - 6 | 711 | 108 | |
GenBank mRNA | AK000505.1 | - | 5 - 6 | 690 | 108 |
AK304885.1 | - | 5 - 6 | 673 | 108 | |
AL136677.1 | - | 5 - 6 | 668 | 108 | |
BC012514.1 | - | 5 - 6 | 662 | 108 |