The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270875.1 | NP_001257804.1 | 1 - 2 | 163 | 59 |
NM_001270876.1 | NP_001257805.1 | 1 - 2 | 163 | 59 | |
NM_022756.5 | NP_073593.2 | 1 - 2 | 163 | 59 | |
NR_073090.1 | 1 - 2 | 163 | 59 | ||
NR_073091.1 | 1 - 2 | 163 | 59 | ||
NR_073092.1 | 1 - 2 | 163 | 59 | ||
GenBank mRNA | AK021792.1 | - | 1 - 2 | 98 | 59 |
AK225434.1 | - | 1 - 2 | 98 | 59 | |
AK225598.1 | - | 1 - 2 | 98 | 59 | |
AK308118.1 | - | 1 - 2 | 98 | 59 | |
AK309663.1 | - | 1 - 2 | 124 | 59 | |
AY007166.1 | - | 1 - 2 | 88 | 59 | |
AY211926.1 | - | 1 - 2 | 87 | 59 | |
BC016328.1 | - | 1 - 2 | 122 | 59 | |
BC056406.1 | - | 1 - 2 | 119 | 59 | |
BM840891.1 | - | 1 - 2 | 107 | 59 | |
BX538212.1 | - | 1 - 2 | 99 | 59 | |
BX640719.1 | - | 1 - 2 | 108 | 59 | |
DQ099384.1 | - | 1 - 2 | 90 | 59 |