Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185147025-185147534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A4
Entrez Gene ID
291
Gene Name
solute carrier family 25 member 4
Gene Aliases
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1
UniGene
Hs.246506
Gene Chromosome Location
Chr.4: 185143263-185150384 on Build GRCh38
Transcripts
NM_001151.3

SNP Detail

SNP ID Chromosome Location
rs914444938 Chr.4: 185147296-185147296 on Build GRCh38
rs1047942863 Chr.4: 185147405-185147405 on Build GRCh38
rs7660552 Chr.4: 185147483-185147483 on Build GRCh38
rs886059268 Chr.4: 185147297-185147299 on Build GRCh38
rs993449716 Chr.4: 185147321-185147321 on Build GRCh38
rs1014106793 Chr.4: 185147148-185147148 on Build GRCh38
rs763730709 Chr.4: 185147059-185147059 on Build GRCh38
rs757138615 Chr.4: 185147345-185147345 on Build GRCh38
rs1027002168 Chr.4: 185147101-185147101 on Build GRCh38
rs562129249 Chr.4: 185147468-185147468 on Build GRCh38
rs955236731 Chr.4: 185147175-185147175 on Build GRCh38
rs1042093401 Chr.4: 185147065-185147065 on Build GRCh38
rs894275865 Chr.4: 185147134-185147134 on Build GRCh38
rs948772204 Chr.4: 185147308-185147308 on Build GRCh38
rs907177789 Chr.4: 185147378-185147378 on Build GRCh38
rs1027932356 Chr.4: 185147166-185147166 on Build GRCh38
rs78882199 Chr.4: 185147452-185147452 on Build GRCh38
rs904551977 Chr.4: 185147083-185147083 on Build GRCh38
rs956143613 Chr.4: 185147464-185147467 on Build GRCh38
rs554157417 Chr.4: 185147430-185147430 on Build GRCh38
rs34935621 Chr.4: 185147308-185147309 on Build GRCh38
rs542187665 Chr.4: 185147096-185147096 on Build GRCh38
rs980156493 Chr.4: 185147498-185147498 on Build GRCh38
rs910306102 Chr.4: 185147407-185147407 on Build GRCh38
rs386683346 Chr.4: 185147482-185147483 on Build GRCh38
rs886059267 Chr.4: 185147233-185147235 on Build GRCh38
rs1045574896 Chr.4: 185147510-185147510 on Build GRCh38
rs986710753 Chr.4: 185147178-185147178 on Build GRCh38
rs34804401 Chr.4: 185147277-185147278 on Build GRCh38
rs547652281 Chr.4: 185147269-185147269 on Build GRCh38
rs951756719 Chr.4: 185147381-185147381 on Build GRCh38
rs555000462 Chr.4: 185147463-185147463 on Build GRCh38
rs542820181 Chr.4: 185147229-185147231 on Build GRCh38
rs146348453 Chr.4: 185147482-185147482 on Build GRCh38
rs767635362 Chr.4: 185147419-185147419 on Build GRCh38
rs371951997 Chr.4: 185147476-185147476 on Build GRCh38
rs112983771 Chr.4: 185147273-185147273 on Build GRCh38
rs190251889 Chr.4: 185147248-185147248 on Build GRCh38
rs1006466096 Chr.4: 185147415-185147415 on Build GRCh38
rs964147811 Chr.4: 185147409-185147409 on Build GRCh38
rs535455404 Chr.4: 185147457-185147457 on Build GRCh38
rs1000660709 Chr.4: 185147106-185147106 on Build GRCh38
rs551018560 Chr.4: 185147463-185147464 on Build GRCh38
rs373871091 Chr.4: 185147122-185147122 on Build GRCh38
rs1804551 Chr.4: 185147104-185147104 on Build GRCh38
rs886059269 Chr.4: 185147395-185147395 on Build GRCh38
rs1055851722 Chr.4: 185147475-185147475 on Build GRCh38
rs755819590 Chr.4: 185147436-185147436 on Build GRCh38
rs886059270 Chr.4: 185147474-185147475 on Build GRCh38
rs922796585 Chr.4: 185147437-185147437 on Build GRCh38
rs139462979 Chr.4: 185147294-185147296 on Build GRCh38
rs112970805 Chr.4: 185147144-185147145 on Build GRCh38
rs375325238 Chr.4: 185147401-185147401 on Build GRCh38
rs570755334 Chr.4: 185147272-185147272 on Build GRCh38

View Full Product Details