Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186198771-186199296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
UniGene
Hs.587231
Gene Chromosome Location
Chr.4: 186191520-186213463 on Build GRCh38
Transcripts
NM_207352.3 XM_005262935.3 XM_017008037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123810 c.683A>G p.E228G Chr.4: 186198965-186198965 on Build GRCh38
4412394 c.676A>G p.M226V Chr.4: 186198958-186198958 on Build GRCh38
733877 c.708G>A p.M236I Chr.4: 186198990-186198990 on Build GRCh38
402546 c.755G>C p.W252S Chr.4: 186199037-186199037 on Build GRCh38
7298653 c.687G>A p.M229I Chr.4: 186198969-186198969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188431476 Chr.4: 186199243-186199243 on Build GRCh38
rs373203551 Chr.4: 186199134-186199134 on Build GRCh38
rs751298422 Chr.4: 186199125-186199125 on Build GRCh38
rs890337525 Chr.4: 186198823-186198823 on Build GRCh38
rs13146537 Chr.4: 186199207-186199207 on Build GRCh38
rs767723431 Chr.4: 186199049-186199050 on Build GRCh38
rs775911832 Chr.4: 186199050-186199050 on Build GRCh38
rs980868011 Chr.4: 186198893-186198893 on Build GRCh38
rs1010979245 Chr.4: 186198870-186198870 on Build GRCh38
rs761190623 Chr.4: 186199054-186199054 on Build GRCh38
rs13146272 Chr.4: 186199057-186199057 on Build GRCh38
rs777920575 Chr.4: 186199133-186199133 on Build GRCh38
rs758339688 Chr.4: 186199120-186199120 on Build GRCh38
rs931773317 Chr.4: 186198803-186198803 on Build GRCh38
rs967091230 Chr.4: 186199186-186199186 on Build GRCh38
rs1030998676 Chr.4: 186198852-186198852 on Build GRCh38
rs759932318 Chr.4: 186198966-186198968 on Build GRCh38
rs779872570 Chr.4: 186198959-186198959 on Build GRCh38
rs769997131 Chr.4: 186198973-186198973 on Build GRCh38
rs775749608 Chr.4: 186199070-186199071 on Build GRCh38
rs372138232 Chr.4: 186199030-186199030 on Build GRCh38
rs969548066 Chr.4: 186198882-186198882 on Build GRCh38
rs761362530 Chr.4: 186199084-186199084 on Build GRCh38
rs375981153 Chr.4: 186199107-186199107 on Build GRCh38
rs756835076 Chr.4: 186198932-186198932 on Build GRCh38
rs926184659 Chr.4: 186198841-186198841 on Build GRCh38
rs950160125 Chr.4: 186198898-186198898 on Build GRCh38
rs977627385 Chr.4: 186198798-186198798 on Build GRCh38
rs894263525 Chr.4: 186199185-186199185 on Build GRCh38
rs764875448 Chr.4: 186199093-186199093 on Build GRCh38
rs992940688 Chr.4: 186199082-186199082 on Build GRCh38
rs184064846 Chr.4: 186199266-186199266 on Build GRCh38
rs369646297 Chr.4: 186199128-186199128 on Build GRCh38
rs369063468 Chr.4: 186198976-186198976 on Build GRCh38
rs372835618 Chr.4: 186199022-186199022 on Build GRCh38
rs531538384 Chr.4: 186199018-186199018 on Build GRCh38
rs991584281 Chr.4: 186198854-186198854 on Build GRCh38
rs180902728 Chr.4: 186199262-186199262 on Build GRCh38
rs758265175 Chr.4: 186198876-186198876 on Build GRCh38
rs199476193 Chr.4: 186199043-186199043 on Build GRCh38
rs35822987 Chr.4: 186199167-186199167 on Build GRCh38
rs1022774043 Chr.4: 186198877-186198877 on Build GRCh38
rs919251190 Chr.4: 186198957-186198957 on Build GRCh38
rs1029768377 Chr.4: 186199148-186199148 on Build GRCh38
rs1047029447 Chr.4: 186198984-186198984 on Build GRCh38
rs774983868 Chr.4: 186198836-186198836 on Build GRCh38
rs543494566 Chr.4: 186198947-186198947 on Build GRCh38
rs778773155 Chr.4: 186198926-186198926 on Build GRCh38
rs899616707 Chr.4: 186199061-186199061 on Build GRCh38
rs199476192 Chr.4: 186199041-186199042 on Build GRCh38
rs753395888 Chr.4: 186198928-186198928 on Build GRCh38
rs183762311 Chr.4: 186198865-186198865 on Build GRCh38
rs13146549 Chr.4: 186199223-186199223 on Build GRCh38
rs72646269 Chr.4: 186199265-186199265 on Build GRCh38
rs771019590 Chr.4: 186199011-186199011 on Build GRCh38
rs763898348 Chr.4: 186198908-186198908 on Build GRCh38
rs940462131 Chr.4: 186199184-186199184 on Build GRCh38
rs1050219918 Chr.4: 186199242-186199242 on Build GRCh38
rs112401237 Chr.4: 186198851-186198851 on Build GRCh38
rs779919994 Chr.4: 186199121-186199121 on Build GRCh38
rs563685672 Chr.4: 186198968-186198968 on Build GRCh38
rs374715498 Chr.4: 186198946-186198946 on Build GRCh38
rs527492432 Chr.4: 186199202-186199202 on Build GRCh38
rs568743864 Chr.4: 186198936-186198936 on Build GRCh38
rs567980520 Chr.4: 186199150-186199150 on Build GRCh38
rs753561422 Chr.4: 186199062-186199062 on Build GRCh38
rs1050351035 Chr.4: 186198810-186198810 on Build GRCh38
rs774674046 Chr.4: 186198959-186198962 on Build GRCh38
rs749306127 Chr.4: 186198977-186198977 on Build GRCh38

View Full Product Details