The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024936.2 | NP_079212.2 | 10 - 11 | 1247 | 101 |
XM_011513835.1 | XP_011512137.1 | 11 - 12 | 1259 | 101 | |
XM_017008128.1 | XP_016863617.1 | 9 - 10 | 1126 | 101 | |
XM_017008129.1 | XP_016863618.1 | 8 - 9 | 962 | 101 | |
GenBank mRNA | AK026677.1 | - | 5 - 6 | 516 | 101 |
AK299211.1 | - | 10 - 11 | 1237 | 101 | |
AY629351.1 | - | 10 - 11 | 1240 | 101 | |
BC016914.1 | - | 7 - 8 | 901 | 101 | |
BC157834.1 | - | 10 - 11 | 1241 | 101 | |
BC171821.1 | - | 10 - 11 | 1241 | 101 |