The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025179.3 | NP_079455.3 | 31 - 32 | 6348 | 78 |
XM_005273164.2 | XP_005273221.1 | 32 - 33 | 6607 | 78 | |
GenBank mRNA | AB007932.1 | - | 31 - 32 | 5799 | 78 |
AK021813.1 | - | 2 - 3 | 531 | 78 | |
AK094367.1 | - | 2 - 3 | 1626 | 78 | |
BC006193.2 | - | 9 - 10 | 1263 | 78 | |
BC009343.2 | - | 14 - 15 | 2338 | 78 | |
BC032125.2 | - | 2 - 3 | 465 | 78 | |
BC132676.1 | - | 31 - 32 | 5703 | 78 | |
BC136530.1 | - | 31 - 32 | 5703 | 78 |