Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLI2
Entrez Gene ID
2736
Gene Name
GLI family zinc finger 2
Gene Aliases
CJS, HPE9, PHS2, THP1, THP2
Chromosome Location
Chr.2: 120797291 - 120992653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005270.4 NP_005261.2 11 - 12 1989 122
XM_006712422.2 XP_006712485.1 11 - 12 1938 122
XM_011510969.2 XP_011509271.2 11 - 12 2621 122
XM_011510970.2 XP_011509272.1 11 - 12 3632 122
XM_011510971.2 XP_011509273.1 11 - 12 2882 122
XM_011510972.2 XP_011509274.2 11 - 12 2019 122
XM_011510973.2 XP_011509275.1 10 - 11 1770 122
XM_011510974.2 XP_011509276.1 10 - 11 1778 122
XM_017003818.1 XP_016859307.1 11 - 12 2570 122
GenBank mRNA AB007295.1 - 8 - 9 1546 122
AB007296.1 - 8 - 9 1495 122
AB007297.1 - 8 - 9 1546 122
AB007298.1 - 8 - 9 1495 122
AB209354.1 - 3 - 4 566 122
BC111410.1 - 11 - 12 1933 122
D14827.1 - 8 - 9 1495 122
D14828.1 - 7 - 8 1391 122
DQ086814.1 - 11 - 12 1959 122

View Full Product Details