Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
S1PR5
Entrez Gene ID
53637
Gene Name
sphingosine-1-phosphate receptor 5
Gene Aliases
EDG8, Edg-8, S1P5, SPPR-1, SPPR-2
Chromosome Location
Chr.19: 10512742 - 10517992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF331840.1 - 1 - 1 607 57
AK074661.1 - 2 - 2 724 57
AK094880.1 - 3 - 3 572 57
BC034703.1 - 2 - 2 655 57
BC067781.1 - 2 - 2 670 57

View Full Product Details