The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030891.4 | NP_112153.1 | 2 - 2 | 709 | 65 |
GenBank mRNA | AB058646.1 | - | 1 - 1 | 402 | 65 |
AY061857.1 | - | 1 - 1 | 402 | 65 | |
AY358120.1 | - | 2 - 2 | 671 | 65 | |
BC064321.1 | - | 2 - 2 | 709 | 65 | |
BC119648.2 | - | 1 - 1 | 499 | 65 | |
BC119649.1 | - | 1 - 1 | 502 | 65 |