The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017440.2 | NP_001017440.1 | 5 - 6 | 1054 | 93 |
NM_031468.3 | NP_113656.2 | 6 - 7 | 1044 | 93 | |
XM_011516594.2 | XP_011514896.1 | 5 - 6 | 896 | 93 | |
XM_011516595.1 | XP_011514897.1 | 5 - 6 | 644 | 93 | |
XM_011516596.2 | XP_011514898.1 | 6 - 7 | 1539 | 93 | |
XM_011516597.2 | XP_011514899.1 | 5 - 6 | 1472 | 93 | |
XM_011516599.1 | XP_011514901.1 | 5 - 6 | 843 | 93 | |
XM_017012675.1 | XP_016868164.1 | 7 - 8 | 1399 | 93 | |
XM_017012676.1 | XP_016868165.1 | 6 - 7 | 1181 | 93 | |
XM_017012677.1 | XP_016868166.1 | 5 - 6 | 694 | 93 | |
XM_017012678.1 | XP_016868167.1 | 6 - 7 | 1156 | 93 | |
XM_017012679.1 | XP_016868168.1 | 5 - 6 | 1509 | 93 | |
XM_017012680.1 | XP_016868169.1 | 5 - 6 | 808 | 93 | |
XM_017012681.1 | XP_016868170.1 | 5 - 6 | 671 | 93 | |
XM_017012682.1 | XP_016868171.1 | 5 - 6 | 1221 | 93 | |
XM_017012683.1 | XP_016868172.1 | 6 - 7 | 1005 | 93 | |
GenBank mRNA | AF282250.1 | - | 6 - 7 | 775 | 93 |
AK123862.1 | - | 5 - 6 | 589 | 93 | |
AK313054.1 | - | 5 - 6 | 644 | 93 | |
AY007302.1 | - | 5 - 6 | 702 | 93 | |
BC020200.1 | - | 5 - 6 | 826 | 93 |