The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031496.1 | NP_113684.1 | 1 - 2 | 2501 | 86 |
GenBank mRNA | BC003126.1 | - | 1 - 2 | 2501 | 86 |