Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGTR1
Entrez Gene ID
185
Gene Name
angiotensin II receptor type 1
Gene Aliases
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
Chromosome Location
Chr.3: 148697871 - 148743003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004835.4 NP_004826.5 2 - 3 312 80
NM_031850.3 NP_114038.4 3 - 4 396 80

View Full Product Details