The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031896.4 | NP_114102.2 | 1 - 2 | 289 | 54 |
XM_005259124.3 | XP_005259181.1 | 2 - 3 | 316 | 54 | |
XM_017027093.1 | XP_016882582.1 | 2 - 3 | 316 | 54 | |
GenBank mRNA | AF288387.1 | - | 1 - 2 | 209 | 54 |
AF361353.1 | - | 1 - 2 | 194 | 54 | |
AF458897.1 | - | 1 - 2 | 194 | 54 | |
BC069332.1 | - | 1 - 2 | 209 | 54 | |
BC093869.1 | - | 1 - 2 | 289 | 54 | |
BC113503.1 | - | 1 - 2 | 289 | 54 |