The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031917.2 | NP_114123.2 | 1 - 2 | 182 | 85 |
XM_011528347.2 | XP_011526649.1 | 2 - 3 | 211 | 85 | |
XM_011528350.2 | XP_011526652.1 | 2 - 3 | 696 | 85 | |
XM_017027347.1 | XP_016882836.1 | 2 - 3 | 369 | 85 | |
GenBank mRNA | AB054064.1 | - | 1 - 2 | 182 | 85 |
AF230330.1 | - | 1 - 2 | 231 | 85 | |
BC142632.1 | - | 1 - 2 | 100 | 85 |