The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018927.3 | NP_061750.1 | 1 - 1 | 1038 | 79 |
NM_032101.2 | NP_115272.1 | 1 - 1 | 1084 | 79 | |
GenBank mRNA | AF152336.1 | - | 1 - 1 | 871 | 79 |
AF152523.1 | - | 1 - 1 | 871 | 79 | |
BC051788.2 | - | 1 - 1 | 1065 | 79 |