Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FGFR1
- Entrez Gene ID
-
2260
- Gene Name
- fibroblast growth factor receptor 1
- Gene Aliases
-
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
- Chromosome Location
-
Chr.8: 38411138 - 38468834 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK024388.1 |
- |
1 - 1 |
1302 |
134 |
View Full Product Details