The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032206.4 | NP_115582.4 | 23 - 24 | 3638 | 104 |
XM_005256193.2 | XP_005256250.1 | 24 - 25 | 3699 | 104 | |
XM_005256194.3 | XP_005256251.1 | 24 - 25 | 3910 | 104 | |
XM_005256195.2 | XP_005256252.1 | 24 - 25 | 3699 | 104 | |
XM_005256196.2 | XP_005256253.1 | 23 - 24 | 3615 | 104 | |
XM_005256197.2 | XP_005256254.1 | 24 - 25 | 3699 | 104 | |
XM_005256199.3 | XP_005256256.1 | 24 - 25 | 3699 | 104 | |
XM_005256201.3 | XP_005256258.1 | 23 - 24 | 3609 | 104 | |
XM_006721297.2 | XP_006721360.1 | 24 - 25 | 3687 | 104 | |
XM_006721298.2 | XP_006721361.1 | 23 - 24 | 3609 | 104 | |
XM_006721300.2 | XP_006721363.1 | 24 - 25 | 3699 | 104 | |
XM_011523373.2 | XP_011521675.1 | 24 - 25 | 3846 | 104 | |
XM_011523375.2 | XP_011521677.1 | 24 - 25 | 3699 | 104 | |
XM_011523376.2 | XP_011521678.1 | 19 - 20 | 1525 | 104 | |
XM_017023766.1 | XP_016879255.1 | 24 - 25 | 3699 | 104 | |
XM_017023767.1 | XP_016879256.1 | 24 - 25 | 3687 | 104 | |
XM_017023768.1 | XP_016879257.1 | 24 - 25 | 3699 | 104 | |
XM_017023769.1 | XP_016879258.1 | 24 - 25 | 3687 | 104 | |
XM_017023770.1 | XP_016879259.1 | 24 - 25 | 3699 | 104 | |
XM_017023771.1 | XP_016879260.1 | 24 - 25 | 3699 | 104 | |
GenBank mRNA | AF389420.1 | - | 22 - 23 | 3511 | 104 |
AK025362.1 | - | 19 - 20 | 2770 | 104 | |
AK027414.1 | - | 4 - 5 | 315 | 104 | |
AK027416.1 | - | 4 - 5 | 314 | 104 | |
AK074133.1 | - | 12 - 13 | 964 | 104 | |
AK090439.1 | - | 19 - 20 | 2693 | 104 | |
BC050527.1 | - | 8 - 9 | 603 | 104 | |
BC063566.1 | - | 6 - 7 | 424 | 104 | |
BX640842.1 | - | 19 - 20 | 2083 | 104 | |
EF452236.1 | - | 22 - 23 | 3511 | 104 |