Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYND12
Entrez Gene ID
84217
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
-
Chromosome Location
Chr.1: 42430329 - 42456267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146192.1 NP_001139664.1 1 - 2 380 67
NM_032257.4 NP_115633.3 1 - 2 380 67
GenBank mRNA AK057384.1 - 1 - 2 380 67
AL136858.1 - 1 - 2 187 67

View Full Product Details