Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOA1
Entrez Gene ID
84273
Gene Name
nitric oxide associated 1
Gene Aliases
C4orf14, MTG3, hAtNOS1, hNOA1, mAtNOS1
Chromosome Location
Chr.4: 56963344 - 56977660 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032313.3 NP_115689.1 5 - 6 1846 65
GenBank mRNA AK074953.1 - 5 - 6 1774 65
AK098215.1 - 6 - 7 1723 65
BC004894.2 - 5 - 6 1846 65

View Full Product Details