The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164750.1 | NP_001158222.1 | 1 - 2 | 176 | 134 |
NM_001164751.1 | NP_001158223.1 | 1 - 2 | 176 | 134 | |
NM_001164752.1 | NP_001158224.1 | 1 - 2 | 176 | 134 | |
NM_001164753.1 | NP_001158225.1 | 1 - 2 | 176 | 134 | |
NM_020164.4 | NP_064549.1 | 1 - 2 | 176 | 134 | |
NM_032467.3 | NP_115856.1 | 1 - 2 | 176 | 134 | |
NM_032468.4 | NP_115857.1 | 1 - 2 | 176 | 134 | |
XM_005251240.1 | XP_005251297.1 | 1 - 2 | 118 | 134 | |
XM_005251244.1 | XP_005251301.1 | 1 - 2 | 118 | 134 | |
XM_017013426.1 | XP_016868915.1 | 1 - 2 | 118 | 134 | |
XM_017013427.1 | XP_016868916.1 | 1 - 2 | 118 | 134 | |
XM_017013435.1 | XP_016868924.1 | 1 - 2 | 118 | 134 | |
XM_017013440.1 | XP_016868929.1 | 1 - 2 | 118 | 134 | |
GenBank mRNA | AF224468.1 | - | 1 - 2 | 126 | 134 |
AF224469.1 | - | 1 - 2 | 101 | 134 | |
AF306765.1 | - | 1 - 2 | 97 | 133 | |
AK295528.1 | - | 1 - 2 | 140 | 135 | |
AK304314.1 | - | 1 - 2 | 132 | 134 | |
BC144362.1 | - | 1 - 2 | 106 | 134 | |
BC144363.1 | - | 1 - 2 | 107 | 135 | |
DN993915.1 | - | 1 - 2 | 184 | 134 |