The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206691.1 | NP_001193620.1 | 16 - 17 | 1966 | 84 |
NM_032491.5 | NP_115880.2 | 12 - 13 | 1632 | 84 | |
NM_213594.2 | NP_998759.1 | 16 - 17 | 2213 | 84 | |
GenBank mRNA | AB095365.1 | - | 17 - 18 | 1961 | 84 |
AB095366.1 | - | 16 - 17 | 2213 | 84 | |
AB195784.1 | - | 7 - 8 | 806 | 84 | |
AB195785.1 | - | 7 - 8 | 728 | 84 | |
AF332192.1 | - | 13 - 14 | 1626 | 84 | |
AK074913.1 | - | 13 - 14 | 1544 | 84 | |
AK291445.1 | - | 16 - 17 | 2198 | 84 | |
AK315698.1 | - | 12 - 13 | 1612 | 84 | |
AL833921.1 | - | 9 - 10 | 1036 | 84 | |
AY102009.1 | - | 16 - 17 | 1866 | 84 | |
BC028582.2 | - | 12 - 13 | 1635 | 84 | |
BC030644.1 | - | 12 - 13 | 1607 | 84 |