The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318124.1 | NP_001305053.1 | 1 - 2 | 348 | 66 |
NM_032562.4 | NP_115951.2 | 1 - 2 | 348 | 66 | |
XM_005270244.4 | XP_005270301.1 | 1 - 2 | 573 | 66 | |
GenBank mRNA | AF339053.1 | - | 1 - 2 | 307 | 66 |
AF349540.1 | - | 1 - 2 | 251 | 66 | |
AK313712.1 | - | 1 - 2 | 311 | 66 | |
AY358032.1 | - | 1 - 2 | 281 | 66 | |
BC093996.1 | - | 1 - 2 | 270 | 66 | |
BC111946.1 | - | 1 - 2 | 270 | 66 | |
BC143532.1 | - | 1 - 2 | 270 | 66 | |
BP274516.1 | - | 1 - 2 | 348 | 66 |