The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256856.1 | NP_001243785.1 | 14 - 14 | 3344 | 86 |
NM_003939.4 | NP_003930.1 | 14 - 14 | 3314 | 86 | |
NM_033637.3 | NP_378663.1 | 15 - 15 | 3422 | 86 | |
XM_006718054.2 | XP_006718117.1 | 15 - 15 | 3722 | 86 | |
XM_011540320.2 | XP_011538622.1 | 12 - 12 | 3158 | 86 | |
XM_017016870.1 | XP_016872359.1 | 14 - 14 | 3644 | 86 | |
XM_017016871.1 | XP_016872360.1 | 15 - 15 | 3313 | 86 | |
XM_017016872.1 | XP_016872361.1 | 15 - 15 | 3485 | 86 | |
XM_017016873.1 | XP_016872362.1 | 15 - 15 | 3341 | 86 | |
XM_017016874.1 | XP_016872363.1 | 13 - 13 | 3236 | 86 | |
GenBank mRNA | BC006204.2 | - | 1 - 1 | 384 | 86 |
CR749295.1 | - | 14 - 14 | 3276 | 86 |