Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17RC
Entrez Gene ID
84818
Gene Name
interleukin 17 receptor C
Gene Aliases
CANDF9, IL17-RL, IL17RL
Chromosome Location
Chr.3: 9917074 - 9933627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203263.1 NP_001190192.1 9 - 10 1061 85
NM_001203264.1 NP_001190193.1 9 - 10 1061 85
NM_001203265.1 NP_001190194.1 8 - 9 1016 85
NM_032732.5 NP_116121.2 8 - 9 1016 85
NM_153460.3 NP_703190.1 9 - 10 1061 85
NM_153461.3 NP_703191.1 9 - 10 1250 85
NR_037807.1 7 - 8 1227 85
XM_017007326.1 XP_016862815.1 9 - 10 1250 85
XM_017007327.1 XP_016862816.1 9 - 10 1250 85
XM_017007328.1 XP_016862817.1 9 - 10 1211 85
XM_017007329.1 XP_016862818.1 9 - 10 1250 85
XM_017007330.1 XP_016862819.1 9 - 10 1211 85
XM_017007331.1 XP_016862820.1 8 - 9 1205 85
XM_017007332.1 XP_016862821.1 8 - 9 1205 85
XM_017007333.1 XP_016862822.1 9 - 10 1250 85
XM_017007334.1 XP_016862823.1 8 - 9 1175 85
XM_017007335.1 XP_016862824.1 8 - 9 1166 85
XM_017007336.1 XP_016862825.1 9 - 10 1250 85
XM_017007337.1 XP_016862826.1 7 - 8 1130 85
XM_017007338.1 XP_016862827.1 8 - 9 1166 85
XM_017007339.1 XP_016862828.1 8 - 9 1205 85
XM_017007340.1 XP_016862829.1 9 - 10 1038 85
XM_017007341.1 XP_016862830.1 8 - 9 993 85
XM_017007342.1 XP_016862831.1 8 - 9 962 85
XM_017007343.1 XP_016862832.1 8 - 9 962 85
XM_017007344.1 XP_016862833.1 9 - 10 999 85
XM_017007345.1 XP_016862834.1 8 - 9 954 85
XM_017007346.1 XP_016862835.1 8 - 9 993 85
XM_017007347.1 XP_016862836.1 7 - 8 917 85
XM_017007348.1 XP_016862837.1 8 - 9 954 85
XM_017007349.1 XP_016862838.1 7 - 8 917 85
GenBank mRNA AF458065.1 - 9 - 10 1016 85
AK075534.1 - 7 - 8 1192 85
AK092907.1 - 9 - 10 1251 85
AK096824.1 - 8 - 9 1158 85
AK225650.1 - 9 - 10 1006 85
AK295377.1 - 7 - 8 1035 85
AK303171.1 - 7 - 8 827 85
AK308241.1 - 8 - 9 946 85
AK315024.1 - 7 - 8 1204 85
AK315053.1 - 8 - 9 961 85
AY358840.1 - 8 - 9 978 85
AY359098.1 - 8 - 9 1006 85
EF676032.1 - 8 - 9 927 85
EF676033.1 - 9 - 10 819 85
EF676034.1 - 9 - 10 830 85

View Full Product Details