The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032773.3 | NP_116162.1 | 4 - 5 | 697 | 97 |
XM_005269362.2 | XP_005269419.1 | 4 - 5 | 1068 | 97 | |
XM_005269363.2 | XP_005269420.1 | 4 - 5 | 1068 | 97 | |
XM_005269365.2 | XP_005269422.1 | 4 - 5 | 1068 | 97 | |
XM_005269367.2 | XP_005269424.1 | 4 - 5 | 1068 | 97 | |
XM_006713791.3 | XP_006713854.1 | 4 - 5 | 1068 | 97 | |
XM_006713792.3 | XP_006713855.1 | 4 - 5 | 1068 | 97 | |
XM_006713793.2 | XP_006713856.1 | 4 - 5 | 1068 | 97 | |
XM_011513242.2 | XP_011511544.1 | 4 - 5 | 1068 | 97 | |
XM_011513243.2 | XP_011511545.1 | 4 - 5 | 1068 | 97 | |
XM_017007350.1 | XP_016862839.1 | 4 - 5 | 1068 | 97 | |
XM_017007351.1 | XP_016862840.1 | 4 - 5 | 1068 | 97 | |
XM_017007352.1 | XP_016862841.1 | 4 - 5 | 1068 | 97 | |
GenBank mRNA | AK293745.1 | - | 4 - 5 | 686 | 97 |
AK303521.1 | - | 4 - 5 | 659 | 97 | |
BC007504.2 | - | 4 - 5 | 649 | 97 | |
DB454969.1 | - | 2 - 3 | 98 |