The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282736.1 | NP_001269665.1 | 6 - 7 | 1369 | 109 |
NM_032812.8 | NP_116201.7 | 7 - 8 | 1516 | 109 | |
XM_011519750.2 | XP_011518052.1 | 7 - 8 | 1744 | 109 | |
GenBank mRNA | AF378757.1 | - | 7 - 8 | 888 | 109 |
AK027529.1 | - | 2 - 3 | 229 | 109 | |
AK304591.1 | - | 8 - 9 | 1400 | 109 | |
AY358486.1 | - | 7 - 8 | 1516 | 109 | |
BC012885.1 | - | 6 - 7 | 975 | 109 |