Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNFT2
Entrez Gene ID
84900
Gene Name
ring finger protein, transmembrane 2
Gene Aliases
TMEM118
Chromosome Location
Chr.12: 116738291 - 116853631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK027533.1 - 3 - 4 274 90

View Full Product Details