Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFYVE19
Entrez Gene ID
84936
Gene Name
zinc finger FYVE-type containing 19
Gene Aliases
ANCHR, MPFYVE
Chromosome Location
Chr.15: 40807076 - 40814569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077268.1 NP_001070736.1 6 - 7 1339 62
NM_001258421.1 NP_001245350.1 6 - 7 1142 62
NM_032850.4 NP_116239.3 7 - 8 1008 62
XM_017022683.1 XP_016878172.1 5 - 6 1901 62
GenBank mRNA AF445414.1 - 7 - 8 850 62
AK027746.1 - 6 - 7 771 62
AK122779.1 - 6 - 7 1129 62
AK225553.1 - 6 - 7 762 62

View Full Product Details