The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193308.1 | NP_001180237.1 | 1 - 2 | 156 | 112 |
NM_032872.2 | NP_116261.1 | 1 - 2 | 156 | 112 | |
XM_006710991.1 | XP_006711054.1 | 1 - 2 | 107 | 112 | |
GenBank mRNA | AK027902.1 | - | 1 - 2 | 126 | 112 |
AY037157.1 | - | 1 - 2 | 83 | 112 | |
BC035725.1 | - | 1 - 2 | 63 | 112 | |
DA435960.1 | - | 1 - 2 | 156 | 112 | |
DA948740.1 | - | 1 - 2 | 133 | 112 |