The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080481.1 | NP_001073950.1 | 5 - 6 | 1011 | 130 |
XM_005267169.3 | XP_005267226.1 | 5 - 6 | 614 | 130 | |
XM_005267170.4 | XP_005267227.1 | 5 - 6 | 583 | 130 | |
XM_005267172.3 | XP_005267229.1 | 5 - 6 | 583 | 130 | |
XM_005267175.3 | XP_005267232.1 | 5 - 6 | 582 | 130 | |
XM_011536200.2 | XP_011534502.1 | 5 - 6 | 1841 | 130 | |
XM_017011382.1 | XP_016866871.1 | 5 - 6 | 1008 | 130 | |
XM_017011383.1 | XP_016866872.1 | 5 - 6 | 534 | 130 | |
XM_017011384.1 | XP_016866873.1 | 5 - 6 | 511 | 130 | |
XM_017011385.1 | XP_016866874.1 | 5 - 6 | 582 | 130 | |
XM_017011386.1 | XP_016866875.1 | 5 - 6 | 582 | 130 | |
GenBank mRNA | AJ583819.2 | - | 4 - 5 | 478 | 130 |
AK310643.1 | - | 5 - 6 | 515 | 130 | |
AL713747.2 | - | 3 - 4 | 377 | 130 | |
BC005991.1 | - | 5 - 6 | 529 | 130 | |
BC150648.2 | - | 5 - 6 | 578 | 130 | |
BC157838.1 | - | 5 - 6 | 578 | 130 |