Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC45A3
Entrez Gene ID
85414
Gene Name
solute carrier family 45 member 3
Gene Aliases
IPCA-2, IPCA-6, IPCA-8, IPCA6, PCANAP2, PCANAP6, PCANAP8, PRST
Chromosome Location
Chr.1: 205657851 - 205680502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033102.2 NP_149093.1 4 - 5 1561 66
XM_005245556.3 XP_005245613.1 4 - 5 1727 66
XM_005245557.4 XP_005245614.1 4 - 5 1653 66
XM_017002638.1 XP_016858127.1 4 - 5 2100 66
GenBank mRNA AK290364.1 - 4 - 5 1562 66
AK313479.1 - 4 - 5 1552 66
AL832933.1 - 2 - 3 625 66
AY033593.1 - 4 - 5 1505 66
BC050416.2 - 4 - 5 1469 66

View Full Product Details