Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALP
Entrez Gene ID
85569
Gene Name
galanin like peptide
Gene Aliases
-
Chromosome Location
Chr.19: 56176020 - 56185775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033106.3 NP_149097.1 2 - 3 174 86
GenBank mRNA AF188492.1 - 2 - 3 174 86
AF188493.1 - 2 - 3 174 86

View Full Product Details