Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- N4BP2L2
- Entrez Gene ID
-
10443
- Gene Name
- NEDD4 binding protein 2 like 2
- Gene Aliases
-
92M18.3, CG005, CG016, PFAAP5
- Chromosome Location
-
Chr.13: 32432488 - 32542710 on Build GRCh38
View Full Product Details