The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033118.3 | NP_149109.1 | 8 - 9 | 1330 | 67 |
GenBank mRNA | AF325549.1 | - | 7 - 8 | 1228 | 67 |
AJ272502.1 | - | 8 - 9 | 1336 | 67 | |
BC007753.1 | - | 1 - 2 | 166 | 67 | |
BC069627.1 | - | 7 - 8 | 1228 | 67 | |
BC092413.1 | - | 8 - 9 | 1330 | 67 | |
BC127622.1 | - | 7 - 8 | 1228 | 67 |