The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014765.1 | NP_001014765.1 | 3 - 4 | 768 | 72 |
NM_001304790.1 | NP_001291719.1 | 3 - 4 | 426 | 72 | |
NM_001304791.1 | NP_001291720.1 | 2 - 3 | 507 | 72 | |
NM_033182.5 | NP_149438.2 | 2 - 3 | 396 | 72 | |
NM_183412.2 | NP_904319.1 | 2 - 3 | 507 | 72 | |
NM_183413.2 | NP_904320.1 | 2 - 3 | 396 | 72 | |
NR_130909.1 | 2 - 3 | 262 | 72 | ||
XM_005263535.1 | XP_005263592.1 | 2 - 3 | 367 | 72 | |
XM_005263536.4 | XP_005263593.1 | 3 - 4 | 761 | 72 | |
XM_005263537.1 | XP_005263594.1 | 2 - 3 | 507 | 72 | |
XM_006711043.2 | XP_006711106.1 | 2 - 3 | 367 | 72 | |
XM_006711045.2 | XP_006711108.1 | 2 - 3 | 369 | 72 | |
XM_011542435.1 | XP_011540737.1 | 3 - 4 | 829 | 72 | |
XM_017002842.1 | XP_016858331.1 | 2 - 3 | 367 | 72 | |
XM_017002843.1 | XP_016858332.1 | 2 - 3 | 369 | 72 | |
XM_017002844.1 | XP_016858333.1 | 2 - 3 | 367 | 72 | |
GenBank mRNA | AK055344.1 | - | 2 - 3 | 507 | 72 |
AK293721.1 | - | 2 - 3 | 344 | 72 | |
AK294053.1 | - | 2 - 3 | 330 | 72 | |
AK296025.1 | - | 2 - 3 | 242 | 72 | |
AK301418.1 | - | 2 - 3 | 376 | 72 | |
AK316284.1 | - | 2 - 3 | 376 | 72 | |
AK316347.1 | - | 2 - 3 | 353 | 72 | |
AY007380.1 | - | 1 - 2 | 303 | 72 | |
AY040878.1 | - | 2 - 3 | 352 | 72 | |
BC007832.2 | - | 2 - 3 | 343 | 72 | |
BM546141.1 | - | 2 - 3 | 357 | 72 | |
BM903606.1 | - | - | 403 | 72 | |
BU164151.1 | - | 3 - 4 | 350 | 72 | |
CR749368.1 | - | 3 - 4 | 757 | 72 | |
HY152948.1 | - | 2 - 3 | 395 | 72 |