The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001311160.1 | NP_001298089.1 | 4 - 4 | 2294 | 99 |
NM_001311162.1 | NP_001298091.1 | 4 - 4 | 1430 | 99 | |
NM_006288.4 | NP_006279.2 | 4 - 4 | 2358 | 99 | |
GenBank mRNA | AF261093.1 | - | 4 - 4 | 1400 | 99 |
AK057865.1 | - | 4 - 4 | 1438 | 99 | |
AK090996.1 | - | 4 - 4 | 2358 | 99 | |
AK094374.1 | - | 5 - 5 | 1368 | 99 | |
AK314018.1 | - | 1 - 1 | 201 | 99 | |
AL161958.1 | - | 4 - 4 | 1373 | 99 |