The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316968.1 | NP_001303897.1 | 2 - 3 | 525 | 119 |
NM_001316969.1 | NP_001303898.1 | 2 - 3 | 241 | 119 | |
NM_033211.3 | NP_149988.1 | 2 - 3 | 285 | 119 | |
GenBank mRNA | AF038182.1 | - | 2 - 3 | 179 | 119 |
AK058020.1 | - | 2 - 3 | 221 | 119 | |
AK090558.1 | - | 2 - 3 | 457 | 119 | |
BC009203.2 | - | 2 - 3 | 246 | 119 | |
BG722102.1 | - | 2 - 3 | 295 | 119 | |
DA503423.1 | - | 2 - 3 | 241 | 119 |