The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033318.4 | NP_201575.3 | 1 - 2 | 267 | 64 |
XM_011530509.2 | XP_011528811.1 | 1 - 2 | 271 | 64 | |
GenBank mRNA | AK312142.1 | - | 1 - 2 | 258 | 64 |
AL449243.1 | - | 1 - 2 | 250 | 64 | |
CR456453.1 | - | 1 - 2 | 207 | 64 |