The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000308.3 | NP_000299.2 | 11 - 12 | 2325 | 65 |
NM_001127695.2 | NP_001121167.1 | 11 - 12 | 2371 | 65 | |
NM_001167594.2 | NP_001161066.1 | 10 - 11 | 2274 | 65 | |
NR_133656.1 | 11 - 12 | 2332 | 65 | ||
GenBank mRNA | AB209705.2 | - | 11 - 12 | 1141 | 65 |
AK097786.1 | - | 10 - 11 | 1038 | 65 | |
AK172808.1 | - | 11 - 12 | 2329 | 65 | |
AK304536.1 | - | 10 - 11 | 1102 | 65 | |
AK307701.1 | - | 11 - 12 | 1114 | 65 | |
AK312898.1 | - | 11 - 12 | 1134 | 65 | |
BC000597.2 | - | 11 - 12 | 1127 | 65 | |
BC093009.1 | - | 11 - 12 | 1134 | 65 | |
M22960.1 | - | 10 - 11 | 1095 | 65 |