The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000739.2 | NP_000730.1 | 4 - 4 | 1246 | 66 |
NM_001006626.1 | NP_001006627.1 | 5 - 5 | 1287 | 66 | |
NM_001006627.1 | NP_001006628.1 | 3 - 3 | 1420 | 66 | |
NM_001006628.1 | NP_001006629.1 | 3 - 3 | 1172 | 66 | |
NM_001006629.1 | NP_001006630.1 | 2 - 2 | 1103 | 66 | |
NM_001006630.1 | NP_001006631.1 | 4 - 4 | 1498 | 66 | |
NM_001006631.1 | NP_001006632.1 | 4 - 4 | 1190 | 66 | |
NM_001006632.1 | NP_001006633.1 | 3 - 3 | 1168 | 66 | |
XM_011515769.2 | XP_011514071.1 | 3 - 3 | 2857 | 66 | |
GenBank mRNA | AF385588.1 | - | 1 - 1 | 875 | 66 |
AF498916.1 | - | 1 - 1 | 875 | 66 | |
AK290236.1 | - | 3 - 3 | 1399 | 66 | |
AL832585.1 | - | 3 - 3 | 1404 | 66 | |
BC095547.1 | - | 1 - 1 | 878 | 66 | |
BC106741.1 | - | 1 - 1 | 875 | 66 | |
BC106742.1 | - | 1 - 1 | 875 | 66 |