Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location
Chr.11: 83455009 - 85628534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142699.1 NP_001136171.1 20 - 21 2428 72
NM_001142700.1 NP_001136172.1 12 - 13 1517 72
NM_001142702.1 NP_001136174.1 3 - 4 559 72
NM_001206769.1 NP_001193698.1 14 - 15 2112 72
NM_001300983.1 NP_001287912.1 15 - 16 2104 72
NM_001364.3 NP_001355.2 15 - 16 2104 72
XM_005273810.4 XP_005273867.1 14 - 15 1747 72
XM_005273811.4 XP_005273868.1 14 - 15 1747 72
XM_011544778.2 XP_011543080.1 21 - 22 3719 72
XM_011544780.2 XP_011543082.1 21 - 22 3719 72
XM_011544782.2 XP_011543084.1 16 - 17 2190 72
XM_011544788.2 XP_011543090.1 13 - 14 1591 72
XM_017017254.1 XP_016872743.1 21 - 22 3719 72
XM_017017255.1 XP_016872744.1 20 - 21 3680 72
XM_017017256.1 XP_016872745.1 20 - 21 3209 72
XM_017017257.1 XP_016872746.1 19 - 20 4141 72
XM_017017258.1 XP_016872747.1 18 - 19 3317 72
XM_017017259.1 XP_016872748.1 19 - 20 3134 72
XM_017017260.1 XP_016872749.1 18 - 19 4066 72
XM_017017261.1 XP_016872750.1 20 - 21 3563 72
XM_017017262.1 XP_016872751.1 14 - 15 2172 72
XM_017017263.1 XP_016872752.1 16 - 17 2190 72
XM_017017264.1 XP_016872753.1 15 - 16 2367 72
XM_017017265.1 XP_016872754.1 15 - 16 2364 72
XM_017017266.1 XP_016872755.1 14 - 15 2170 72
XM_017017267.1 XP_016872756.1 15 - 16 2272 72
XM_017017268.1 XP_016872757.1 15 - 16 2093 72
XM_017017269.1 XP_016872758.1 15 - 16 2794 72
XM_017017270.1 XP_016872759.1 16 - 17 2190 72
XM_017017271.1 XP_016872760.1 15 - 16 2367 72
XM_017017272.1 XP_016872761.1 15 - 16 2364 72
XM_017017273.1 XP_016872762.1 13 - 14 2023 72
XM_017017274.1 XP_016872763.1 15 - 16 2272 72
XM_017017275.1 XP_016872764.1 13 - 14 2015 72
XM_017017276.1 XP_016872765.1 15 - 16 2792 72
XM_017017277.1 XP_016872766.1 14 - 15 2016 72
XM_017017278.1 XP_016872767.1 13 - 14 2016 72
XM_017017279.1 XP_016872768.1 15 - 16 2034 72
XM_017017280.1 XP_016872769.1 15 - 16 2711 72
XM_017017281.1 XP_016872770.1 14 - 15 2211 72
XM_017017282.1 XP_016872771.1 14 - 15 1747 72
XM_017017283.1 XP_016872772.1 14 - 15 2016 72
XM_017017284.1 XP_016872773.1 13 - 14 1592 72
XM_017017285.1 XP_016872774.1 14 - 15 2638 72
XM_017017286.1 XP_016872775.1 11 - 12 1456 72
XM_017017287.1 XP_016872776.1 4 - 5 476 72
XM_017017288.1 XP_016872777.1 3 - 4 315 72
XM_017017289.1 XP_016872778.1 3 - 4 1890 72
XM_017017290.1 XP_016872779.1 3 - 4 535 72
XM_017017291.1 XP_016872780.1 3 - 4 378 72
XM_017017292.1 XP_016872781.1 3 - 4 1890 72
GenBank mRNA AK126776.1 - 12 - 13 1517 72
AK294375.1 - 14 - 15 1635 72
AK295094.1 - 15 - 16 2104 72
AK295624.1 - 14 - 15 1727 72
AK299778.1 - 3 - 4 528 72
AK299797.1 - 15 - 16 1862 72
CR749820.1 - 20 - 21 2428 72
CR933674.1 - 3 - 4 528 72
KJ535093.1 - 20 - 21 2311 72
U32376.1 - 15 - 16 2077 72

View Full Product Details