The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314039.1 | NP_001300968.1 | 1 - 2 | 181 | 106 |
NM_001539.3 | NP_001530.1 | 1 - 2 | 181 | 106 | |
GenBank mRNA | AK289623.1 | - | 1 - 2 | 109 | 106 |
AK298725.1 | - | 1 - 2 | 109 | 106 | |
BC008182.1 | - | 1 - 2 | 94 | 106 | |
BI462555.1 | - | 1 - 2 | 176 | 106 | |
BM469197.1 | - | 1 - 2 | 82 | 106 | |
D13388.1 | - | 1 - 2 | 107 | 106 | |
DA003255.1 | - | 1 - 2 | 183 | 107 | |
L08069.1 | - | 1 - 2 | 72 | 105 |