Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL5
Entrez Gene ID
4636
Gene Name
myosin light chain 5
Gene Aliases
-
Chromosome Location
Chr.4: 674199 - 682033 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002477.1 NP_002468.1 1 - 2 106 59
XM_017008245.1 XP_016863734.1 2 - 3 1784 59
XM_017008246.1 XP_016863735.1 3 - 4 1653 59
GenBank mRNA L03785.1 - 1 - 2 106 59

View Full Product Details