Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRM
Entrez Gene ID
5797
Gene Name
protein tyrosine phosphatase, receptor type M
Gene Aliases
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location
Chr.18: 7567316 - 8406861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105244.1 NP_001098714.1 4 - 5 1054 64
NM_002845.3 NP_002836.3 4 - 5 1054 64
XM_011525708.2 XP_011524010.1 4 - 5 1049 64
XM_011525710.2 XP_011524012.1 4 - 5 1049 64
XM_011525712.2 XP_011524014.1 4 - 5 1049 64
XM_011525713.2 XP_011524015.1 4 - 5 649 64
XM_017025894.1 XP_016881383.1 4 - 5 593 64
XM_017025895.1 XP_016881384.1 4 - 5 593 64
XM_017025896.1 XP_016881385.1 4 - 5 593 64
XM_017025897.1 XP_016881386.1 4 - 5 593 64
XM_017025898.1 XP_016881387.1 4 - 5 593 64
XM_017025899.1 XP_016881388.1 4 - 5 593 64
XM_017025900.1 XP_016881389.1 4 - 5 593 64
XM_017025901.1 XP_016881390.1 4 - 5 593 64
XM_017025902.1 XP_016881391.1 4 - 5 593 64
XM_017025903.1 XP_016881392.1 4 - 5 593 64
XM_017025904.1 XP_016881393.1 4 - 5 593 64
XM_017025905.1 XP_016881394.1 4 - 5 649 64
XM_017025910.1 XP_016881399.1 4 - 5 593 64
XM_017025911.1 XP_016881400.1 4 - 5 1049 64
XM_017025912.1 XP_016881401.1 4 - 5 593 64
GenBank mRNA AK303652.1 - 4 - 5 552 64
BC051651.1 - 4 - 5 790 64
BC151842.1 - 4 - 5 1600 64
X58288.1 - 4 - 5 551 64

View Full Product Details