The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278622.1 | NP_001265551.1 | 3 - 4 | 547 | 82 |
NM_002873.1 | NP_002864.1 | 2 - 3 | 652 | 82 | |
NM_133338.2 | NP_579916.1 | 4 - 5 | 548 | 82 | |
NM_133342.2 | NP_579920.1 | 3 - 4 | 525 | 82 | |
NM_133343.1 | NP_579921.1 | 3 - 4 | 490 | 82 | |
NM_133344.2 | NP_579922.1 | 2 - 3 | 307 | 82 | |
XM_017009679.1 | XP_016865168.1 | 3 - 4 | 367 | 82 | |
XM_017009680.1 | XP_016865169.1 | 2 - 3 | 421 | 82 | |
XM_017009681.1 | XP_016865170.1 | 2 - 3 | 279 | 82 | |
GenBank mRNA | AF017748.1 | - | 2 - 3 | 277 | 82 |
AF076838.1 | - | 2 - 3 | 652 | 82 | |
AF112263.1 | - | 1 - 2 | 129 | 82 | |
AF126424.1 | - | 4 - 5 | 547 | 82 | |
AJ001642.1 | - | 2 - 3 | 316 | 82 | |
AJ004977.1 | - | 2 - 3 | 312 | 82 | |
AK292487.1 | - | 3 - 4 | 525 | 82 | |
AL122068.1 | - | 3 - 4 | 490 | 82 | |
BC032304.1 | - | 1 - 2 | 118 | 82 | |
BX537441.1 | - | 3 - 4 | 547 | 82 |