The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310121.1 | NP_001297050.1 | 10 - 11 | 1222 | 82 |
NM_001310123.1 | NP_001297052.1 | 8 - 9 | 1113 | 82 | |
NM_002973.3 | NP_002964.3 | 10 - 11 | 2016 | 82 | |
NR_132311.1 | 10 - 11 | 2016 | 82 | ||
GenBank mRNA | AK307803.1 | - | 10 - 11 | 1447 | 82 |
BC111757.1 | - | 10 - 11 | 1222 | 82 | |
BC114546.1 | - | 8 - 9 | 1113 | 82 | |
U70323.1 | - | 10 - 11 | 2013 | 82 | |
Y08262.1 | - | 10 - 11 | 1613 | 82 |