The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003064.3 | NP_003055.1 | 1 - 2 | 109 | 69 |
GenBank mRNA | AF114471.1 | - | 1 - 2 | 88 | 69 |
AK312192.1 | - | 1 - 2 | 93 | 69 | |
AX772818.1 | - | 1 - 2 | 109 | 69 | |
BC020708.1 | - | 1 - 2 | 109 | 69 | |
X04470.1 | - | 1 - 2 | 106 | 69 | |
X04503.1 | - | 1 - 2 | 88 | 69 |