Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF43
Entrez Gene ID
7594
Gene Name
zinc finger protein 43
Gene Aliases
HTF6, KOX27, ZNF39L1
Chromosome Location
Chr.19: 21804946 - 21852095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256650.1 NP_001243579.1 1 - 2 164 57
XM_017027210.1 XP_016882699.1 1 - 2 100 57
XM_017027215.1 XP_016882704.1 1 - 2 156 57
XM_017027216.1 XP_016882705.1 1 - 2 124 57
GenBank mRNA X59244.1 - 1 - 2 64 57

View Full Product Details