The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244580.1 | NP_001231509.1 | 13 - 14 | 1329 | 58 |
NM_003496.3 | NP_003487.1 | 13 - 14 | 1329 | 58 | |
GenBank mRNA | AB209489.1 | - | 3 - 4 | 264 | 58 |
AF076974.1 | - | 13 - 14 | 1329 | 58 | |
AF110377.1 | - | 13 - 14 | 1249 | 58 |